Anti-MTHFSD Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-80463
102216-572EA 564.74 USD
102216-572
Anti-MTHFSD Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal MTHFSD Antibody. Tested Applications: Western Blot. Tested Reactivity: Human, Mouse, Bovine, Chicken, Xenopus, Zebrafish.

Type: Primary
Antigen: MTHFSD
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR