Anti-MTHFD2 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-54655
102184-278EA 564.74 USD
102184-278
Anti-MTHFD2 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal MTHFD2 Antibody. Tested Applications: Western Blot. Tested Reactivity: Human, Mouse, Rat, Canine.

Type: Primary
Antigen: MTHFD2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR