Anti-MTERF4 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-98416
102223-148EA 564.74 USD
102223-148
Anti-MTERF4 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal MTERFD2 Antibody. Tested Applications: Western Blot, Immunoblotting. Tested Reactivity: Human, Mouse.

Type: Primary
Antigen: MTERF4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR