Anti-MTCH2 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-54790
102184-542EA 564.74 USD
102184-542
Anti-MTCH2 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal MTCH2 Antibody. Tested Applications: Western Blot. Tested Reactivity: Human, Mouse, Rat.

Type: Primary
Antigen: MTCH2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR