|
![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | MSTO1 |
Antigen Name | Misato 1, Mitochondrial Distribution And Morphology Regulator |
Reactivity | Human |
Conjugation | Unconjugated |
Clonality | Polyclonal |
Antibody Type | Primary |
Host | Rabbit |
Isotype | IgG |
Western Blot | Yes |
ImmunoChemistry | Yes |
Antigen Synonyms | LST005, RP11-29H23.3, protein misato homolog 1, DKFZp686B1757, MST, FLJ10504, DKFZp686I01261, misato homolog 1 (Drosophila) |
Gene ID | 55154 |
Concentration | 0.46 mg/ml |
Immunogen | Recombinant protein encompassing a sequence within the center region of human MSTO1. The exact sequence is proprietary. |
Purification | Immunogen affinity purified |
Storage Buffer | 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol |
Storage Temperature | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |