Anti-MRTO4 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-53203
102183-862EA 564.74 USD
102183-862
Anti-MRTO4 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal MRTO4 Antibody. Tested Applications: Western Blot. Tested Reactivity: Human, Mouse, Rat.

Type: Primary
Antigen: MRTO4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR