Anti-MRPS36 Rabbit Polyclonal Antibody

NBP1-51379
102181-526EA 466.15 USD
102181-526
Anti-MRPS36 Rabbit Polyclonal Antibody
Rabbit Polyclonal MRPS36 Antibody. Tested Applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:50-1:200

Type: Primary
Antigen: MRPS36
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR