Anti-MRPS34 Rabbit Polyclonal Antibody

NBP1-30523
102163-622EA 466.15 USD
102163-622
Anti-MRPS34 Rabbit Polyclonal Antibody
Rabbit Polyclonal MRPS34 Antibody. Tested Applications: Western Blot. Tested Reactivity: Human, Mouse.

Type: Primary
Antigen: MRPS34
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR