Rabbit Polyclonal anti-MRG-1 Antibody, Novus Biologicals (35530002)

35530002
102103-638EA 749.23 USD
102103-638
Rabbit Polyclonal anti-MRG-1 Antibody, Novus Biologicals (35530002)
Rabbit Polyclonal MRG-1 Antibody. Tested Applications: ELISA, Immunocytochemistry/Immunofluorescence. Tested Reactivity: C. elegans.

Type: Primary
Antigen: MRG-1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: C. elegans
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR