|
![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | Mre11 | |
Antigen Name | Meiotic Recombination 11 | |
Reactivity | Rat, Mouse, Chicken, Hamster, Human | Rat, Hamster, Human, Chicken, Mouse |
Conjugation | Unconjugated | |
Clonality | Polyclonal | |
Antibody Type | Primary | |
Host | Rabbit | |
Isotype | IgG | |
Western Blot | Yes | |
ImmunoChemistry | Yes | |
ImmunoPrecipitation | Yes | |
ImmunoFluorescence | Yes | |
Flow Cytometry | Yes | |
Antigen Synonyms | AT-like disease, double-strand break repair protein MRE11A, Meiotic recombination 11 homolog 1, DNA recombination and repair protein, MRE11 homolog A, MRE11 homolog 1, ATLDmeiotic recombination (S. cerevisiae) 11 homolog A, MRE11 meiotic recombination 11 homolog A (S. cerevisiae), HNGS1, MRE11MRE11B, Meiotic recombination 11 homolog A, endo/exonuclease Mre11 | AT-like disease, endo/exonuclease Mre11, Meiotic recombination 11 homolog 1, MRE11 meiotic recombination 11 homolog A (S. cerevisiae), MRE11 homolog A, DNA recombination and repair protein, double-strand break repair protein MRE11A, MRE11 homolog 1, Meiotic recombination 11 homolog A, ATLDmeiotic recombination (S. cerevisiae) 11 homolog A, MRE11MRE11B, HNGS1 |
Gene ID | 4361 | |
Immunogen | Full length human Mre11 protein. [UniProt# P49959] | |
Purification | Unpurified | |
Storage Buffer | Whole antisera | |
Storage Temperature | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |