Anti-CDC42BPA Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-46823
102176-532EA 635.51 USD
102176-532
Anti-CDC42BPA Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal MRCK Antibody. Tested Applications: Immunoprecipitation, Western Blot (-). Tested Reactivity: Human.

Type: Primary
Antigen: CDC42BPA
Clonality: Polyclonal
Clone: 0
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR