Anti-MPPE1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-69305
102198-846EA 564.74 USD
102198-846
Anti-MPPE1 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal MPPE1 Antibody. Tested Applications: Western Blot. Tested Reactivity: Human, Mouse.

Type: Primary
Antigen: MPPE1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR