Anti-MPND Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-79263
102214-172EA 564.74 USD
102214-172
Anti-MPND Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal MPND Antibody. Tested Applications: Western Blot. Tested Reactivity: Human, Canine.

Type: Primary
Antigen: MPND
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR