Anti-MOR42-3 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB110-75065
102125-176EA 564.74 USD
102125-176
Anti-MOR42-3 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal MOR42-3 Antibody. Tested Applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen, Immunohistochemistry-Paraffin. Tested Reactivity: Mouse.

Type: Primary
Antigen: MOR42-3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR