Anti-Monopolin Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB500-131
102131-824EA 417.23 USD
102131-824
Anti-Monopolin Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal Monopolin Antibody. Tested Applications: Western Blot. Tested Reactivity: Yeast.

Type: Primary
Antigen: Monopolin
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Yeast
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR