Anti-MAOB Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-59569
102192-830EA 564.74 USD
102192-830
Anti-MAOB Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal Monoamine Oxidase B Antibody. Tested Applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human, Mouse, Rat, Bovine, Canine, Porcine.

Type: Primary
Antigen: MAOB
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR