Anti-MMP28 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-33768
102167-420EA 349 USD
102167-420
Anti-MMP28 Rabbit Polyclonal Antibody
Rabbit Polyclonal MMP28 Antibody. Tested Applications: Western Blot, Immunocytochemistry/Immunofluorescence. Tested Reactivity: Human.

Type: Primary
Antigen: MMP28
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR