Anti-MMP23B Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-60036
102193-748EA 564.74 USD
102193-748
Anti-MMP23B Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal MMP23B Antibody. Tested Applications: Western Blot. Tested Reactivity: Human, Mouse, Rat.

Type: Primary
Antigen: MMP23B
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR