Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MMP2 Rabbit Polyclonal Antibody (Biotin)

Supplier: Novus Biologicals
NB200-193B
Rabbit Polyclonal MMP2 Antibody [Biotin]. Tested Applications: Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Frozen, Immunohistochemistry-Paraffin. Tested Reactivity: Human, Mouse, Rat, Chicken.

Type: Primary
Antigen: MMP2
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 μl NB200-193B 102129-264 Each Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol MMP-2
Antigen Name Matrix Metalloproteinase 2
Reactivity Human, Rat, Mouse, Guinea Pig, Chicken
Conjugation Biotin
Clonality Polyclonal
Antibody Type Primary
Host Rabbit
Isotype IgG
Western Blot Yes
ELISA Yes
ImmunoChemistry Yes
ImmunoFluorescence Yes
Antigen Synonyms neutrophil gelatinase, TBE-1matrix metalloproteinase 2 (gelatinase A, 72kDa type IVcollagenase), EC 3.4.24, 72kD type IVcollagenase), matrix metalloproteinase-II, matrix metallopeptidase 2 (gelatinase A, 72kDa gelatinase, EC 3.4.24.24, MMP-II, Matrix metalloproteinase-2, CLG4, Gelatinase A, 72kD gelatinase, MMP-2, 72 kDa gelatinase, matrix metalloproteinase 2 (gelatinase A, collagenase type IV-A, CLG4A72 kDa type IV collagenase, MONA
Gene ID 4313
Immunogen A synthetic peptide made to an internal region of the human MMP2 protein (between residues 400-500) [UniProt P08253]
Purification Immunogen affinity purified
Storage Buffer PBS
Storage Temperature Store at 4C in the dark.
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty