Anti-MMP19 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-57092
102189-096EA 564.74 USD
102189-096
Anti-MMP19 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal MMP19 Antibody. Tested Applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human, Canine.

Type: Primary
Antigen: MMP19
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR