Anti-MMP19 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB600-1518
102133-438EA 263 USD
102133-438
Anti-MMP19 Rabbit Polyclonal Antibody
Rabbit Polyclonal MMP19 Antibody. Tested Applications: Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human.

Type: Primary
Antigen: MMP19
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR