Anti-MLL5 Rabbit Polyclonal Antibody

NBP1-02511
102155-416EA 536.92 USD
102155-416
Anti-MLL5 Rabbit Polyclonal Antibody
Rabbit Polyclonal MLL5 Antibody. Tested Applications: ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human.

Recommended Dilutions: Immunohistochemistry (Parafin): 10 ?g/ml

Type: Primary
Antigen: MLL5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR