Anti-MLKL Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-56729
102188-378EA 663.82 USD
102188-378
Anti-MLKL Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal MLKL Antibody. Tested Applications: Western Blot. Tested Reactivity: Human, Rat, Bovine.

Type: Primary
Antigen: MLKL
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR