Anti-MLK3 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-58913
102191-548EA 564.74 USD
102191-548
Anti-MLK3 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal MLK3 Antibody. Tested Applications: Western Blot. Tested Reactivity: Human, Mouse, Rat.

Type: Primary
Antigen: MLK3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR