Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-KIF23 Rabbit Polyclonal Antibody (Agarose immobilized)

Supplier: Novus Biologicals
NB600-422
Rabbit Polyclonal MKLP1 Antibody [Agarose Immobilized]. Tested Applications: Immunoprecipitation. Tested Reactivity: Human.

Type: Primary
Antigen: KIF23
Clonality: Polyclonal
Clone:
Conjugation: Agarose immobilized
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 μg NB600-422 102133-944 Each Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol MKLP1
Antigen Name Kinesin Family Member 23
Reactivity Human
Conjugation Agarose Immobilized
Clonality Polyclonal
Antibody Type Primary
Host Rabbit
Isotype IgG
ImmunoPrecipitation Yes
Antigen Synonyms MKLP1Mitotic kinesin-like protein 1, MKLP-1, kinesin-like 5 (mitotic kinesin-like protein 1), KNSL5CHO1, kinesin-like protein KIF23, Kinesin-like protein 5, mitotic kinesin-like 1, kinesin family member 23
Gene ID 9493
Immunogen The immunogen recognized by this antibody maps to a region between residues 1 and 50 of human Mitotic Kinesin-Like 1 using the numbering given in entry NP_004847.2 (GeneID 9493).
Purification Immunogen affinity purified
Storage Buffer PBS
Storage Temperature Store at 4C. Do not freeze.
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping