Anti-MFN1 Rabbit Polyclonal Antibody (Biotin)

Supplier: Novus Biologicals
NBP1-51841B
102182-284EA 614.15 USD
102182-284
Anti-MFN1 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Rabbit Polyclonal Mitofusin-1 Antibody [Biotin]. Tested Applications: Immunocytochemistry/Immunofluorescence. Tested Reactivity: Human.

Type: Primary
Antigen: MFN1
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR