Anti-Mitofilin Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB100-1919SS NB100-1919
102106-348EA 244.92 USD
102106-348 102106-346
Anti-Mitofilin Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal Mitofilin Antibody. Tested Applications: Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human, Mouse, Rat, Canine, Feline.

Type: Primary
Antigen: Mitofilin
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR