Anti-MIS18A Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-70753
102200-104EA 564.74 USD
102200-104
Anti-MIS18A Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal MIS18A Antibody. Tested Applications: Western Blot. Tested Reactivity: Human.

Type: Primary
Antigen: MIS18A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR