Anti-MIS12 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB100-2358
102107-208EA 635.51 USD
102107-208
Anti-MIS12 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal MIS12 Antibody. Tested Applications: Immunoprecipitation, Western Blot (-). Tested Reactivity: Human.

Type: Primary
Antigen: MIS12
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR