|
![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | MIPEP |
Antigen Name | Mitochondrial Intermediate Peptidase |
Reactivity | Human |
Conjugation | Unconjugated |
Clonality | Polyclonal |
Antibody Type | Primary |
Host | Rabbit |
Isotype | IgG |
Western Blot | Yes |
ImmunoChemistry | Yes |
ImmunoFluorescence | Yes |
Antigen Synonyms | MIPEC 3.4.24.59, mitochondrial intermediate peptidase, HMIP |
Gene ID | 4285 |
Concentration | 0.53 mg/ml |
Immunogen | Recombinant protein encompassing a sequence within the center region of human MIPEP. The exact sequence is proprietary. |
Purification | Immunogen affinity purified |
Storage Buffer | 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol |
Storage Temperature | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |