Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CCL4/MIP-1 beta Rabbit Polyclonal Antibody (Biotin) [clone: ]

Supplier: Novus Biologicals
NBP1-77722
The CCL4 / MIP-1 beta Antibody [Biotin] from Novus Biologicals is a rabbit polyclonal antibody to CCL4 / MIP-1 beta. This antibody reacts with human. The CCL4 / MIP-1 beta Antibody [Biotin] has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: CCL4/MIP-1 beta
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 μg NBP1-77722 102211-932 Each Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Shipping duration cannot exceed 2 days.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol CCL4/MIP-1 beta
Antigen Name Chemokine (C-C motif) Ligand 4
Reactivity Human
Conjugation Biotin
Clonality Polyclonal
Antibody Type Primary
Host Rabbit
Isotype IgG
Western Blot Yes
ELISA Yes
Antigen Synonyms G-26, secreted protein G-26, chemokine (C-C motif) ligand 4, MIP1B, Protein H400, MIP1B1, G-26 T-lymphocyte-secreted protein, SCYA2, HC21, small inducible cytokine A4 (homologous to mouse Mip-1b), MIP-1-beta(1-69), Lymphocyte activation gene 1 protein, LAG-1, Macrophage inflammatory protein 1-beta, AT744.1, MIP-1-beta, PAT 744, C-C motif chemokine 4, SIS-gamma, MIP-1 beta/CCL4, ACT-2, ACT2, Small-inducible cytokine A4, SCYA4, T-cell activation protein 2, LAG1
Gene ID 6351
Immunogen This purified antibody was prepared from whole rabbit serum produced by repeated immunizations with full length recombinant human MIP1 beta protein.
Purification Delipidation and Defibrination
Storage Buffer 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free
Storage Temperature Store at 4C. Do not freeze.
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty