Anti-MIF4GD Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-57562
102190-006EA 564.74 USD
102190-006
Anti-MIF4GD Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal MIF4GD Antibody. Tested Applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human, Mouse, Rat, Canine.

Type: Primary
Antigen: MIF4GD
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR