Anti-MIF Rabbit Polyclonal Antibody

NBP2-13035
102252-850EA 466.15 USD
102252-850
Anti-MIF Rabbit Polyclonal Antibody
Rabbit Polyclonal MIF Antibody. Tested Applications: Western Blot. Tested Reactivity: Human, Mouse, Rat.

Type: Primary
Antigen: MIF
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR