Anti-MIF Rabbit Polyclonal Antibody

NB100-1910
102106-328EA 352.31 USD
102106-328
Anti-MIF Rabbit Polyclonal Antibody
Rabbit Polyclonal MIF Antibody. Tested Applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunoprecipitation. Tested Reactivity: Mouse, Rat.

Type: Primary
Antigen: MIF
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR