Anti-MK Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-31158
102164-226EA 379 USD
102164-226
Anti-MK Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal Midkine Antibody. Tested Applications: Western Blot, Dot Blot. Tested Reactivity: Human.

Type: Primary
Antigen: MK
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR