Anti-MICALL1 Rabbit Polyclonal Antibody

NBP1-79460
102214-568EA 466.15 USD
102214-568
Anti-MICALL1 Rabbit Polyclonal Antibody
Rabbit Polyclonal MICALL1 Antibody. Tested Applications: Western Blot. Tested Reactivity: Human.

Type: Primary
Antigen: MICALL1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR