Anti-MGP Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-57998
102190-694EA 239 USD
102190-694
Anti-MGP Rabbit Polyclonal Antibody
Rabbit Polyclonal MGP Antibody. Tested Applications: Western Blot. Tested Reactivity: Human.

Type: Primary
Antigen: MGP
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR