|
![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | GRM6/GRM7 |
Antigen Name | Glutamate Receptor 6/7 |
Reactivity | Human |
Conjugation | Unconjugated |
Clonality | Polyclonal |
Antibody Type | Primary |
Host | Rabbit |
Isotype | IgG |
Western Blot | Yes |
ImmunoChemistry | Yes |
Antigen Synonyms | GluR6/7, Glutamate receptor 6, Glutamate receptor 7, Excitatory amino acid receptor 5, GluR-7, GRIK3, EAA5, GluR-6, GluR6, GRIK2, EAA4, Excitatory amino acid receptor 4, GluR7 |
Gene ID | 2898 |
Concentration | 0.2 mg/ml |
Immunogen | Synthetic peptide from C-terminus of human GluR6/7. |
Purification | Immunogen affinity purified |
Storage Buffer | PBS (pH 7.6) and 1.0% BSA |
Storage Temperature | Store at 4C. Do not freeze. |