|
![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | MGC4172 |
Reactivity | Human |
Conjugation | Unconjugated |
Clonality | Polyclonal |
Antibody Type | Primary |
Host | Rabbit |
Isotype | IgG |
Western Blot | Yes |
Antigen Synonyms | short chain dehydrogenase/reductase family 24C, ARPG836, dehydrogenase/reductase (SDR family) member 11, member 1, dehydrogenase/reductase SDR family member 11, SDR24C1, MGC4172, short-chain dehydrogenase/reductase, FLJ39232 |
Gene ID | 79154 |
Immunogen | Synthetic peptides corresponding to MGC4172(short-chain dehydrogenase/reductase) The peptide sequence was selected from the middle region of MGC4172. Peptide sequence DGHIININSMSGHRVLPLSVTHFYSATKYAVTALTEGLRQELREAQTHIR. |
Purification | Immunogen affinity purified |
Storage Buffer | PBS and 2% Sucrose |
Storage Temperature | Store at -20C. Avoid freeze-thaw cycles. |