Anti-MGC13138 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-79709
102215-066EA 564.74 USD
102215-066
Anti-MGC13138 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal MGC13138 Antibody. Tested Applications: Western Blot. Tested Reactivity: Human.

Type: Primary
Antigen: MGC13138
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR