Anti-MFAP5 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-77942
102212-260EA 777.05 USD
102212-260
Anti-MFAP5 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal MFAP5 Antibody. Tested Applications: Western Blot, ELISA. Tested Reactivity: Human, Mouse, Rat, Bovine, Primate.

Type: Primary
Antigen: MFAP5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR