Anti-MFAP3L Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-70637
102199-884EA 564.74 USD
102199-884
Anti-MFAP3L Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal MFAP3L Antibody. Tested Applications: Western Blot. Tested Reactivity: Human, Mouse, Rat, Canine.

Type: Primary
Antigen: MFAP3L
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR