Anti-MEX3D Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-77332
102211-502EA 564.74 USD
102211-502
Anti-MEX3D Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal MEX3D Antibody. Tested Applications: Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human.

Type: Primary
Antigen: MEX3D
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR