Anti-METT10D Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-79256
102214-158EA 564.74 USD
102214-158
Anti-METT10D Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal METT10D Antibody. Tested Applications: Western Blot. Tested Reactivity: Human, Mouse, Rat, Bovine, Canine, Chicken, Xenopus, Zebrafish.

Type: Primary
Antigen: METT10D
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR