Anti-GRM6 Rabbit Polyclonal Antibody

NBP1-31088
102164-120EA 493.85 USD
102164-120
Anti-GRM6 Rabbit Polyclonal Antibody
Rabbit Polyclonal metabotropic Glutamate Receptor 6 Antibody. Tested Applications: Western Blot, Dot Blot. Tested Reactivity: Human.

Type: Primary
Antigen: metabotropic Glutamate Receptor 6
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR