Anti-MGLUR5/MGLUR1A Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB300-126
102129-964EA 491.08 USD
102129-964
Anti-MGLUR5/MGLUR1A Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal metabotropic Glutamate Receptor 5/1 Antibody. Tested Applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen. Tested Reactivity: Rat.

Type: Primary
Antigen: MGLUR5/MGLUR1A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR