Anti-GRM2/GRM3 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB300-124
102129-962EA 329 USD
102129-962
Anti-GRM2/GRM3 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal metabotropic Glutamate Receptor 2/3 Antibody. Tested Applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen. Tested Reactivity: Rat.

Type: Primary
Antigen: metabotropic Glutamate Receptor 2/3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR