Anti-MET Rabbit Polyclonal Antibody

NBP1-51450
102181-660EA 466.15 USD
102181-660
Anti-MET Rabbit Polyclonal Antibody
Rabbit Polyclonal Met (c-Met) Antibody. Tested Applications: Western Blot. Tested Reactivity: Human, Mouse, Rat.

Type: Primary
Antigen: MET
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR