Anti-MET Rabbit Polyclonal Antibody

NBP1356801.0ML NBP1356800.5ML NBP1356807.0ML NBP1356800.1ML
102167-850EA 466.15 USD
102167-850 102167-852 102167-854 102167-848
Anti-MET Rabbit Polyclonal Antibody
Rabbit Polyclonal Met (c-Met) Antibody. Tested Applications: Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:50

Type: Primary
Antigen: MET
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR