Rabbit Polyclonal anti-mes-3 Antibody, Novus Biologicals (38580002)

38580002
102103-810EA 749.23 USD
102103-810
Rabbit Polyclonal anti-mes-3 Antibody, Novus Biologicals (38580002)
Rabbit Polyclonal mes-3 Antibody. Tested Applications: ELISA, Immunocytochemistry/Immunofluorescence. Tested Reactivity: C. elegans.

Type: Primary
Antigen: mes-3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: C. elegans
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR